Alfa Aesar™ 3-(Benzylamino)propionitrile, 97+%

Manufacturer: Alfa Aesar™ 4194906

Quantity 5g

 View more versions of this product

Catalog No. AA4194906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 706-03-6
    Formula Weight 160.22
    MDL Number MFCD00001955
    UN Number UN3276
    Chemical Name or Material 3-(Benzylamino)propionitrile

    Specifications

    Formula Weight 160.22
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.5308
    MDL Number MFCD00001955
    Flash Point >110° (230°F)
    Physical Form Liquid
    UN Number UN3276
    Density 1.024g/mL
    Health Hazard 1 Danger
    Quantity 5g
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    CAS 706-03-6
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Chemical Name or Material 3-(Benzylamino)propionitrile
    Assay ≥97%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit