Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-(Benzylamino)propylamine, 95%

Click to view available options
Quantity:
250mg
1g
5g
13910-48-0
C10H16N2
164.252
MFCD00274361
RFLHDXQRFPJPRR-UHFFFAOYSA-N
n1-benzylpropane-1,3-diamine,3-benzylamino propylamine,n-benzylpropane-1,3-diamine,3-aminopropyl benzyl amine,n-benzyl-1,3-propanediamine,n-3-aminopropyl-n-benzylamine,3-aminopropyl benzylamine,n-benzylpropan-1,3-diamin,n-benzyltrimethylenediamine,n-3-aminopropyl benzylamine
83811
N'-benzylpropane-1,3-diamine
C1=CC=C(C=C1)CNCCCN
13910-48-0
C10H16N2
164.252
MFCD00274361
RFLHDXQRFPJPRR-UHFFFAOYSA-N
n1-benzylpropane-1,3-diamine,3-benzylamino propylamine,n-benzylpropane-1,3-diamine,3-aminopropyl benzyl amine,n-benzyl-1,3-propanediamine,n-3-aminopropyl-n-benzylamine,3-aminopropyl benzylamine,n-benzylpropan-1,3-diamin,n-benzyltrimethylenediamine,n-3-aminopropyl benzylamine
83811
N'-benzylpropane-1,3-diamine
C1=CC=C(C=C1)CNCCCN
Refractive Index 1.5320
Color Yellow
Quantity 250mg
Beilstein 509316
Formula Weight 164.25
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 3-(Benzylamino)propylamine