Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxybenzeneboronic acid, 98%, Acros Organics™

Click to view available options
Quantity:
500mg
2.5g
25g
156682-54-1
C13H13BO3
228.054
WIJNYNBSPQMJGO-UHFFFAOYSA-N
3-benzyloxyphenylboronic acid,3-benzyloxybenzeneboronic acid,3-benzyloxy phenyl boronic acid,3-benzyloxy phenylboronic acid,m-benzyloxyphenylboronic acid,3-benzyloxyphenyl boronic acid,3-boronophenoxy methyl benzene,3-benzyl-oxy phenyl boranediol,boronic acid, 3-phenylmethoxy phenyl
2734313
(3-phenylmethoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
156682-54-1
C13H13BO3
228.054
WIJNYNBSPQMJGO-UHFFFAOYSA-N
3-benzyloxyphenylboronic acid,3-benzyloxybenzeneboronic acid,3-benzyloxy phenyl boronic acid,3-benzyloxy phenylboronic acid,m-benzyloxyphenylboronic acid,3-benzyloxyphenyl boronic acid,3-boronophenoxy methyl benzene,3-benzyl-oxy phenyl boranediol,boronic acid, 3-phenylmethoxy phenyl
2734313
(3-phenylmethoxyphenyl)boronic acid
B(C1=CC(=CC=C1)OCC2=CC=CC=C2)(O)O
Quantity 25g
Color White to Beige
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 228.05
Physical Form Crystalline Powder or Crystals
Percent Purity ≥97.5%
Chemical Name or Material 3-(Benzyloxy)phenylboronic acid

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.