Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Benzyloxyacetophenone, 97%

Click to view available options
Quantity:
5g
25g
100g
34068-01-4
C15H14O2
226.275
MFCD00026221
FGQMEAWGAUALJQ-UHFFFAOYSA-N
1-3-benzyloxy phenyl ethanone,3-benzyloxyacetophenone,3'-benzyloxyacetophenone,1-3-benzyloxy phenyl ethan-1-one,1-3-phenylmethoxy phenyl ethan-1-one,3'-benzyloxy acetophenone,ethanone, 1-3-phenylmethoxy phenyl,1-acetyl-3-phenylmethoxy benzene
98689
1-(3-phenylmethoxyphenyl)ethanone
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
34068-01-4
C15H14O2
226.275
MFCD00026221
FGQMEAWGAUALJQ-UHFFFAOYSA-N
1-3-benzyloxy phenyl ethanone,3-benzyloxyacetophenone,3'-benzyloxyacetophenone,1-3-benzyloxy phenyl ethan-1-one,1-3-phenylmethoxy phenyl ethan-1-one,3'-benzyloxy acetophenone,ethanone, 1-3-phenylmethoxy phenyl,1-acetyl-3-phenylmethoxy benzene
98689
1-(3-phenylmethoxyphenyl)ethanone
CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2
Quantity 5g
Formula Weight 226.28
Percent Purity 97%
Chemical Name or Material 3'-Benzyloxyacetophenone