Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3'-Benzyloxyacetophenone, 97%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Quantity | 100g |
Formula Weight | 226.28 |
Percent Purity | 97% |
Chemical Name or Material | 3'-Benzyloxyacetophenone |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 251-816-0
TSCA : No
Recommended Storage : Ambient temperatures