Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Benzyloxyaniline, 98%, Acros Organics

Click to view available options
Quantity:
5g
10g
Packaging:
Glass bottle
1484-26-0
C13H13NO
199.253
MFCD00007784
IGPFOKFDBICQMC-UHFFFAOYSA-N
3-benzyloxyaniline,3-benzyloxy aniline,benzenamine, 3-phenylmethoxy,unii-iab1vda972,3-benzyloxy-phenylamine,iab1vda972,3-phenylmethyl oxy aniline,3-phenylmethoxy phenylamine,3-benzyloxy benzenamine,3-benyloxyaniline
92892
3-phenylmethoxyaniline
C1=CC=C(C=C1)COC2=CC=CC(=C2)N
1484-26-0
C13H13NO
199.253
MFCD00007784
IGPFOKFDBICQMC-UHFFFAOYSA-N
3-benzyloxyaniline,3-benzyloxy aniline,benzenamine, 3-phenylmethoxy,unii-iab1vda972,3-benzyloxy-phenylamine,iab1vda972,3-phenylmethyl oxy aniline,3-phenylmethoxy phenylamine,3-benzyloxy benzenamine,3-benyloxyaniline
92892
3-phenylmethoxyaniline
C1=CC=C(C=C1)COC2=CC=CC(=C2)N
Quantity 10g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige to Tan
Melting Point 62°C to 66°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5CH2OC6H4NH2
Beilstein 13, 404
Formula Weight 199.25
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 3-Benzyloxyaniline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning