Alfa Aesar™ 3-Benzyloxybenzoyl chloride, 95%

Manufacturer: Alfa Aesar™ H5041003

Quantity 1g

 View more versions of this product

Catalog No. AAH5041003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61535-46-4
    Molecular Formula C14H11ClO2
    Formula Weight 246.7
    MDL Number MFCD02258040
    UN Number UN3261
    Chemical Name or Material 3-Benzyloxybenzoyl chloride

    Specifications

    Formula Weight 246.7
    Molecular Formula C14H11ClO2
    Melting Point 40° to 42°C
    MDL Number MFCD02258040
    UN Number UN3261
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    CAS 61535-46-4
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 3-Benzyloxybenzoyl chloride
    Assay 95%

    Description

    Applications
    Reactant for preparation of dihydroquinolinones and tetrahydroquinazolinones as potent kinesin spindle protein inhibitors and potential antitumor agents, preparation of carbamate derivatives as fatty acid amide hydrolase inhibitors.

    Solubility
    Reacts with water.

    Notes
    Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit