Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Biphenylcarboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
716-76-7
C13H10O2
198.22
MFCD00045846
XNLWJFYYOIRPIO-UHFFFAOYSA-N
biphenyl-3-carboxylic acid,3-biphenylcarboxylic acid,1,1'-biphenyl-3-carboxylic acid,diphenyl-3-carboxylic acid,wln: qvr cr,3-biphenylcarboxylicacid,m-phenylbenzoic acid,3-phenyl benzoic acid
12854
3-phenylbenzoic acid
C1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O
716-76-7
C13H10O2
198.22
MFCD00045846
XNLWJFYYOIRPIO-UHFFFAOYSA-N
biphenyl-3-carboxylic acid,3-biphenylcarboxylic acid,1,1'-biphenyl-3-carboxylic acid,diphenyl-3-carboxylic acid,wln: qvr cr,3-biphenylcarboxylicacid,m-phenylbenzoic acid,3-phenyl benzoic acid
12854
3-phenylbenzoic acid
C1=CC=C(C=C1)C2=CC(=CC=C2)C(=O)O
Melting Point 164°C to 169°C
Packaging Glass bottle
Color White to Beige
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5C6H4COOH
Beilstein 09, IV, 2478
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone,diethyl ether,hexane,soluble in benzene,petroleum ether,alkali
Formula Weight 198.22
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-Biphenylcarboxylic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning