Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(BOC-Amino)piperidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
172603-05-3
C10H20N2O2
200.28
WUOQXNWMYLFAHT-UHFFFAOYSA-N
3-boc-aminopiperidine,tert-butyl piperidin-3-ylcarbamate,3-n-boc-aminopiperidine,tert-butyl n-piperidin-3-yl carbamate,3-boc-amino piperidine,3-n-boc-amino piperidine,3-tert-butoxycarbonylamino piperidine,3-aminopiperidine, 3-boc protected,piperidin-3-yl-carbamic acid tert-butyl ester,tert-butyl n-3-piperidyl carbamate
4233041
tert-butyl N-piperidin-3-ylcarbamate
CC(C)(C)OC(=O)NC1CCCNC1
172603-05-3
C10H20N2O2
200.28
WUOQXNWMYLFAHT-UHFFFAOYSA-N
3-boc-aminopiperidine,tert-butyl piperidin-3-ylcarbamate,3-n-boc-aminopiperidine,tert-butyl n-piperidin-3-yl carbamate,3-boc-amino piperidine,3-n-boc-amino piperidine,3-tert-butoxycarbonylamino piperidine,3-aminopiperidine, 3-boc protected,piperidin-3-yl-carbamic acid tert-butyl ester,tert-butyl n-3-piperidyl carbamate
4233041
tert-butyl N-piperidin-3-ylcarbamate
CC(C)(C)OC(=O)NC1CCCNC1
Packaging Glass bottle
Color White
Quantity 5g
Infrared Spectrum Authentic
Melting Point 104.6°C to 112.6°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 200.28
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 3-(BOC-Amino)piperidine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning