Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Bromo-1,2-propanediol, 97%, ACROS Organics™
Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
Specifications
Specific Gravity | 1.77 |
Color | Colorless to Yellow |
Boiling Point | 72.0°C to 75.0°C (0.2mmHg) |
Flash Point | >110°C |
Quantity | 25mL |
Packaging | Glass bottle |
Density | 1.7700g/mL |
Infrared Spectrum | Authentic |
Refractive Index | 1.5165 to 1.5195 |
CAS Min % | 96.0 |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 225-186-2