Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-1,2-propanediol, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
4704-77-2
C3H7BrO2
154.99
MFCD00004711
SIBFQOUHOCRXDL-UHFFFAOYSA-N
3-bromo-1,2-propanediol,monobromoglycerol,alpha-bromohydrin,1,2-propanediol, 3-bromo,bromodeoxyglycerol,glycerol bromohydrin,1-bromo-2,3-propanediol,3-bromodeoxyglycerol,rs-3-bromo-1,2-propanediol,1,2-propanediol,3-bromo
20827
CHEBI:18719
3-bromopropane-1,2-diol
C(C(CBr)O)O
4704-77-2
C3H7BrO2
154.99
MFCD00004711
SIBFQOUHOCRXDL-UHFFFAOYSA-N
3-bromo-1,2-propanediol,monobromoglycerol,alpha-bromohydrin,1,2-propanediol, 3-bromo,bromodeoxyglycerol,glycerol bromohydrin,1-bromo-2,3-propanediol,3-bromodeoxyglycerol,rs-3-bromo-1,2-propanediol,1,2-propanediol,3-bromo
20827
CHEBI:18719
3-bromopropane-1,2-diol
C(C(CBr)O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Density 1.7700g/mL
Refractive Index 1.5165 to 1.5195
Flash Point >110°C
Specific Gravity 1.77
Color Colorless to Yellow
Quantity 100mL
Boiling Point 72.0°C to 75.0°C (0.2mmHg)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula BrCH2CH(OH)CH2OH
Beilstein 01, I, 248
Formula Weight 154.99
Physical Form Viscous Liquid
Percent Purity 97%
Chemical Name or Material 3-Bromo-1, 2-propanediol
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 225-186-2