Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-1-methylpyrrolidine, 97%

Click to view available options
Quantity:
250mg
1g
5g
10603-45-9
C5H10BrN
164.046
MFCD12755877
ZHUYTHOKJZSZQJ-UHFFFAOYSA-N
3-bromo-1-methyl-pyrrolidine,pyrrolidine, 3-bromo-1-methyl,3-bromo-1-methylpyrrolidine
13726279
3-bromo-1-methylpyrrolidine
CN1CCC(C1)Br
10603-45-9
C5H10BrN
164.046
MFCD12755877
ZHUYTHOKJZSZQJ-UHFFFAOYSA-N
3-bromo-1-methyl-pyrrolidine,pyrrolidine, 3-bromo-1-methyl,3-bromo-1-methylpyrrolidine
13726279
3-bromo-1-methylpyrrolidine
CN1CCC(C1)Br
Sensitivity Moisture Sensitive
Quantity 250mg
Color Yellow to Brown
Density 1.361g/mL
Refractive Index 1.4910
UN Number UN2733
Formula Weight 164.04
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Bromo-1-methylpyrrolidine

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

storageNote1 : 2° to 8°C