Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-1-trimethylsilyl-1-propyne, 98%

Manufacturer:  Alfa Aesar™ H5345114

Quantity 25g

 View more versions of this product

Catalog No. AAH5345114


  Check Availability
Add to cart

Specifications

Description

Applications
3-Bromo-1-trimethylsilyl-1-propyne is used in the preparation of tetrahydroisoquinoline-3-carboxylic acid derivatives. It acts as a propargylating agent and used for the alkylation of dianion of beta-keto esters at gamma-carbon. It plays an important role in the synthesis of allenic alcohols and terminal conjugated enynes.

Solubility
Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

38002-45-8
BrCH2C≡CSi(CH3)3
MFCD00134460
3-(Trimethylsilyl)propargyl bromide
C6H11BrSi
191.14
2037650
3-Bromo-1-trimethylsilyl-1-propyne

Specifications

38002-45-8
BrCH2C≡CSi(CH3)3
MFCD00134460
3-(Trimethylsilyl)propargyl bromide
25g
3-Bromo-1-trimethylsilyl-1-propyne
C6H11BrSi
191.14
2037650
44° to 45°C (2mmHg)
63°C (145°F)
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit