Alfa Aesar™ 3-Bromo-2-butanone, 97%, stab. with <1% magnesium oxide

Manufacturer: Alfa Aesar™ A1018630

Quantity 250g

 View more versions of this product

Catalog No. AAA1018630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 814-75-5
    Molecular Formula C4H7BrO
    Formula Weight 151.01
    MDL Number MFCD00013538
    UN Number UN2922
    Beilstein 906773
    Chemical Name or Material 3-Bromo-2-butanone, Stabilized with <1% magnesium oxide

    Specifications

    Formula Weight 151.01
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Quantity 250g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
    Molecular Formula C4H7BrO
    UN Number UN2922
    Boiling Point 69° to 70°C (70mmHg)
    Health Hazard 1 Danger
    TSCA TSCA
    MDL Number MFCD00013538
    CAS 814-75-5
    Beilstein 906773
    Flash Point >100°C (212°F)
    Odor Pungent
    Health Hazard 2 GHS H Statement
    H301-H331-H314-H318-H312
    Toxic if swallowed.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful in contact with skin.
    RTECS Number EL7005000
    Chemical Name or Material 3-Bromo-2-butanone, Stabilized with <1% magnesium oxide
    Assay 97%

    Description

    Applications
    3-Bromo-2-butanone is involved in the preparation of o-isopropyl S-3-oxobutan-2-yl dithiocarbonate by reaction with potassium o-isopropylxanthate, which on treatment with sulfuric acid gives 4,5-dimethyl-1,3-dithiol-2-one .

    Solubility
    Miscible with dichloromethane.

    Notes
    Incompatible with strong oxidizing agents, strong bases and strong reducing agents.

    GHS H Statement
    H301-H331-H314-H318-H312
    Toxic if swallowed.
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful in contact with skin.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit