Learn More
3-Bromo-2-butanone, 97%, stab. with <1% magnesium oxide, Thermo Scientific Chemicals
Description
3-Bromo-2-butanone is involved in the preparation of o-isopropyl S-3-oxobutan-2-yl dithiocarbonate by reaction with potassium o-isopropylxanthate, which on treatment with sulfuric acid gives 4,5-dimethyl-1,3-dithiol-2-one .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Bromo-2-butanone is involved in the preparation of o-isopropyl S-3-oxobutan-2-yl dithiocarbonate by reaction with potassium o-isopropylxanthate, which on treatment with sulfuric acid gives 4,5-dimethyl-1,3-dithiol-2-one .
Solubility
Miscible with dichloromethane.
Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
Density | 1.438 |
Boiling Point | 69°C to 70°C (70 mmHg) |
Flash Point | >100°C (212°F) |
Odor | Pungent |
Refractive Index | 1.459 |
Quantity | 10 g |
UN Number | UN2922 |
Beilstein | 906773 |
Solubility Information | Miscible with dichloromethane. |
Formula Weight | 151.01 |
Show More |
Safety and Handling
Hazard Category | H226-H302+H312+H332-H314 |
Hazard Statement | GHS H Statement H301-H331-H314-H318-H312 Toxic if swallowed. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful in contact with skin. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S. |
EINECSNumber | 212-404-6 |
RTECSNumber | EL7005000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.