Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-2-chloro-5-(trifluoromethyl)pyridine, 97%

Click to view available options
Quantity:
1g
5g
25g
71701-92-3
C6H2BrClF3N
260.438
MFCD09878432
GVQLMGPWTHAUPV-UHFFFAOYSA-N
3-bromo-2-chloro-5-trifluoromethyl pyridine,2-chloro-3-bromo-5-trifluoromethylpyridine,abbypharma ap-13-5058,pyridine, 3-bromo-2-chloro-5-trifluoromethyl,pubchem15158,acmc-1bcir,ksc496i9b,2-morpholin-4-ylethanesulfonicacid,hydrate,3-bromo-2-chloro-5-trifluoromethyl pyridine,2-chloro-3-bromo-5-trifluoromethyl pyridine
13536061
3-bromo-2-chloro-5-(trifluoromethyl)pyridine
C1=C(C=NC(=C1Br)Cl)C(F)(F)F
71701-92-3
C6H2BrClF3N
260.438
MFCD09878432
GVQLMGPWTHAUPV-UHFFFAOYSA-N
3-bromo-2-chloro-5-trifluoromethyl pyridine,2-chloro-3-bromo-5-trifluoromethylpyridine,abbypharma ap-13-5058,pyridine, 3-bromo-2-chloro-5-trifluoromethyl,pubchem15158,acmc-1bcir,ksc496i9b,2-morpholin-4-ylethanesulfonicacid,hydrate,3-bromo-2-chloro-5-trifluoromethyl pyridine,2-chloro-3-bromo-5-trifluoromethyl pyridine
13536061
3-bromo-2-chloro-5-(trifluoromethyl)pyridine
C1=C(C=NC(=C1Br)Cl)C(F)(F)F
Melting Point 28°C to 32°C
Color Brown to Yellow
Quantity 1g
UN Number UN2811
Formula Weight 260.44
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-chloro-5-(trifluoromethyl)pyridine

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.