Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-chloropyridine, 98%, Acros Organics™

Click to view available options
Quantity:
5g
25g
52200-48-3
C5H3BrClN
192.44
HDYNIWBNWMFBDO-UHFFFAOYSA-N
2-chloro-3-bromopyridine,pyridine, 3-bromo-2-chloro,3-bromo-2-chloro-pyridine,pubchem1098,2-chloro-bromopyridine,acmc-209kyb,3-bromo-2-chlorolpyridine,3-bromo-2-chloropyridine,3-bromo-2-chloro pyridine,2-chloro-3-bromo pyridine
693324
3-bromo-2-chloropyridine
C1=CC(=C(N=C1)Cl)Br
52200-48-3
C5H3BrClN
192.44
HDYNIWBNWMFBDO-UHFFFAOYSA-N
2-chloro-3-bromopyridine,pyridine, 3-bromo-2-chloro,3-bromo-2-chloro-pyridine,pubchem1098,2-chloro-bromopyridine,acmc-209kyb,3-bromo-2-chlorolpyridine,3-bromo-2-chloropyridine,3-bromo-2-chloro pyridine,2-chloro-3-bromo pyridine
693324
3-bromo-2-chloropyridine
C1=CC(=C(N=C1)Cl)Br
Boiling Point 97°C (1.3 kPa)
Color White to Yellow
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 57.9°C to 58.3°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 192.44
Physical Form Crystalline Powder, Crystals or Flakes or Solid
Percent Purity 98%
Chemical Name or Material 3-Bromo-2-chloropyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.