Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-2-cyanobenzeneboronic acid, 96%

Click to view available options
Quantity:
250mg
1g
1032231-32-5
C7H5BBrNO2
225.836
LHGIIQFOQAPPQA-UHFFFAOYSA-N
3-bromo-2-cyanophenyl boronic acid,2-cyano-3-bromophenylboronic acid
46739320
(3-bromo-2-cyanophenyl)boronic acid
B(C1=C(C(=CC=C1)Br)C#N)(O)O
1032231-32-5
C7H5BBrNO2
225.836
LHGIIQFOQAPPQA-UHFFFAOYSA-N
3-bromo-2-cyanophenyl boronic acid,2-cyano-3-bromophenylboronic acid
46739320
(3-bromo-2-cyanophenyl)boronic acid
B(C1=C(C(=CC=C1)Br)C#N)(O)O
Quantity 250mg
Formula Weight 225.84
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 3-Bromo-2-cyanobenzeneboronic acid

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures