Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine, 97%

Click to view available options
Quantity:
250mg
1g
5g
1031929-01-7
C6H2BrF4N
243.987
MFCD08061304
HKACHYLUXHBYMO-UHFFFAOYSA-N
3-bromo-2-fluoro-5-trifluoromethyl pyridine,2-fluoro-3-bromo-5-trifluoromethylpyridine,pyridine, 3-bromo-2-fluoro-5-trifluoromethyl,3-bromo-2-fluoro-5-trifluoromethyl ryridine,pubchem15149,pyridine,3-bromo-2-fluoro-5-trifluoromethyl,3-bromo-2-fluoro-5-trifluoromethyl pyridine
23053237
3-bromo-2-fluoro-5-(trifluoromethyl)pyridine
C1=C(C=NC(=C1Br)F)C(F)(F)F
1031929-01-7
C6H2BrF4N
243.987
MFCD08061304
HKACHYLUXHBYMO-UHFFFAOYSA-N
3-bromo-2-fluoro-5-trifluoromethyl pyridine,2-fluoro-3-bromo-5-trifluoromethylpyridine,pyridine, 3-bromo-2-fluoro-5-trifluoromethyl,3-bromo-2-fluoro-5-trifluoromethyl ryridine,pubchem15149,pyridine,3-bromo-2-fluoro-5-trifluoromethyl,3-bromo-2-fluoro-5-trifluoromethyl pyridine
23053237
3-bromo-2-fluoro-5-(trifluoromethyl)pyridine
C1=C(C=NC(=C1Br)F)C(F)(F)F
Refractive Index 1.4600
Quantity 5g
Density 1.774g/mL
UN Number UN1992
Formula Weight 243.99
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.