Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-2-fluorobenzaldehyde, 97%

Click to view available options
Quantity:
5g
25g
100g
149947-15-9
C7H4BrFO
203.01
MFCD09027089
OUAZPCKRSSEQKB-UHFFFAOYSA-N
3-bromo-2-fluorobenzenecarbaldehyde,3-bromo-2-fluorobenzadehyde,2-fluoro-3-bromobenzaldehyde,benzaldehyde,3-bromo-2-fluoro,benzaldehyde, 3-bromo-2-fluoro,3-bromofluorobenzaldehyde,intermediates-zcf02145,acmc-209d3i,3-bromo-2-fluoro-benzaldehyde,3-bromanyl-2-fluoranyl-benzaldehyde
22019361
3-bromo-2-fluorobenzaldehyde
C1=CC(=C(C(=C1)Br)F)C=O
149947-15-9
C7H4BrFO
203.01
MFCD09027089
OUAZPCKRSSEQKB-UHFFFAOYSA-N
3-bromo-2-fluorobenzenecarbaldehyde,3-bromo-2-fluorobenzadehyde,2-fluoro-3-bromobenzaldehyde,benzaldehyde,3-bromo-2-fluoro,benzaldehyde, 3-bromo-2-fluoro,3-bromofluorobenzaldehyde,intermediates-zcf02145,acmc-209d3i,3-bromo-2-fluoro-benzaldehyde,3-bromanyl-2-fluoranyl-benzaldehyde
22019361
3-bromo-2-fluorobenzaldehyde
C1=CC(=C(C(=C1)Br)F)C=O
Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Quantity 25g
Color White
Melting Point 43°C to 46°C
Sensitivity Light Sensitive
Beilstein 13904499
Formula Weight 203.01
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-fluorobenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C