Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-2-fluorobenzeneboronic acid, 98%

Click to view available options
Quantity:
1g
5g
352535-97-8
C6H5BBrFO2
218.816
MFCD05664228
QCLOSORNRRMFIA-UHFFFAOYSA-N
3-bromo-2-fluorophenyl boronic acid,2-fluoro-3-bromophenylboronic acid,boronic acid,b-3-bromo-2-fluorophenyl,boronic acid, b-3-bromo-2-fluorophenyl,3-bromo-2-fluorophenylboronicacid,acmc-1aijv,3-bromo-2-fluoro-phenylboronic acid,3-bromo-2-fluorobenzeneboronic acid,3-bromanyl-2-fluoranyl-phenyl boronic acid,contains varying amounts of anhydride
16217170
(3-bromo-2-fluorophenyl)boronic acid
B(C1=C(C(=CC=C1)Br)F)(O)O
352535-97-8
C6H5BBrFO2
218.816
MFCD05664228
QCLOSORNRRMFIA-UHFFFAOYSA-N
3-bromo-2-fluorophenyl boronic acid,2-fluoro-3-bromophenylboronic acid,boronic acid,b-3-bromo-2-fluorophenyl,boronic acid, b-3-bromo-2-fluorophenyl,3-bromo-2-fluorophenylboronicacid,acmc-1aijv,3-bromo-2-fluoro-phenylboronic acid,3-bromo-2-fluorobenzeneboronic acid,3-bromanyl-2-fluoranyl-phenyl boronic acid,contains varying amounts of anhydride
16217170
(3-bromo-2-fluorophenyl)boronic acid
B(C1=C(C(=CC=C1)Br)F)(O)O
Quantity 5g
Melting Point 225°C to 227°C (literature)
Beilstein 14006009
Formula Weight 218.82
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 3-Bromo-2-fluorobenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning