Alfa Aesar™ 3-Bromo-2-fluorobenzyl bromide, 97%

Manufacturer: Alfa Aesar™ H3222803

Quantity 1g

 View more versions of this product

Catalog No. AAH3222803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 149947-16-0
    Molecular Formula C7H5Br2F
    Formula Weight 267.92
    MDL Number MFCD09842442
    UN Number UN3265
    Chemical Name or Material 3-Bromo-2-fluorobenzyl bromide

    Specifications

    Formula Weight 267.92
    Molecular Formula C7H5Br2F
    MDL Number MFCD09842442
    UN Number UN3265
    Boiling Point 60°C (0.1mmHg)
    Quantity 1g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    CAS 149947-16-0
    Chemical Name or Material 3-Bromo-2-fluorobenzyl bromide
    Assay 97%

    Description

    Applications
    Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit