Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-2-hydroxybenzonitrile, 97%

Click to view available options
Quantity:
1g
5g
13073-28-4
C7H4BrNO
198.019
MFCD11042854
BUWXNBPLRLEXOZ-UHFFFAOYSA-N
2-bromo-6-cyanophenol,benzonitrile,3-bromo-2-hydroxy,acmc-1c4pd,2-hydroxy-3-bromobenzonitrile,3-bromo-2-hydroxy-benzonitrile,benzonitrile, 3-bromo-2-hydroxy,3-bromo-2-hydroxybenznitrile,3-bromo-2-hydroxybenzonitrile,-8-hydroxy-2-dipropylamino tetralin hydrobromide
12468680
3-bromo-2-hydroxybenzonitrile
C1=CC(=C(C(=C1)Br)O)C#N
13073-28-4
C7H4BrNO
198.019
MFCD11042854
BUWXNBPLRLEXOZ-UHFFFAOYSA-N
2-bromo-6-cyanophenol,benzonitrile,3-bromo-2-hydroxy,acmc-1c4pd,2-hydroxy-3-bromobenzonitrile,3-bromo-2-hydroxy-benzonitrile,benzonitrile, 3-bromo-2-hydroxy,3-bromo-2-hydroxybenznitrile,3-bromo-2-hydroxybenzonitrile,-8-hydroxy-2-dipropylamino tetralin hydrobromide
12468680
3-bromo-2-hydroxybenzonitrile
C1=CC(=C(C(=C1)Br)O)C#N
Quantity 5g
Melting Point 118°C to 122°C
Color Brown to White
UN Number UN3439
Formula Weight 198.02
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-hydroxybenzonitrile

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.