Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine, 98%

Click to view available options
Quantity:
5g
25g
124432-63-9
C7H5BrF3NO
256.022
MFCD09972202
YUDYUCKEWCVXTE-UHFFFAOYSA-N
3-bromo-2-methoxy-5-trifluoromethyl pyridine,bromides,acmc-209as0,pyridine,3-bromo-2-methoxy-5-trifluoromethyl,2-methoxy-3-bromo-5-trifluoromethylpyridine,3-bromo-5-trifluoromethyl-2-methoxypyridine
14837018
3-bromo-2-methoxy-5-(trifluoromethyl)pyridine
COC1=C(C=C(C=N1)C(F)(F)F)Br
124432-63-9
C7H5BrF3NO
256.022
MFCD09972202
YUDYUCKEWCVXTE-UHFFFAOYSA-N
3-bromo-2-methoxy-5-trifluoromethyl pyridine,bromides,acmc-209as0,pyridine,3-bromo-2-methoxy-5-trifluoromethyl,2-methoxy-3-bromo-5-trifluoromethylpyridine,3-bromo-5-trifluoromethyl-2-methoxypyridine
14837018
3-bromo-2-methoxy-5-(trifluoromethyl)pyridine
COC1=C(C=C(C=N1)C(F)(F)F)Br
Color White
Quantity 5g
Beilstein 13639286
Formula Weight 256.02
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 3-Bromo-2-methoxy-5-(trifluoromethyl)pyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures