Alfa Aesar™ 3-Bromo-2-methoxypyridine, 95%

Manufacturer: Alfa Aesar™ H2727906

Quantity 5g

 View more versions of this product

Catalog No. AAH2727906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13472-59-8
    Molecular Formula C6H6BrNO
    Formula Weight 188.02
    MDL Number MFCD03095349
    Chemical Name or Material 3-Bromo-2-methoxypyridine

    Specifications

    Formula Weight 188.02
    Molecular Formula C6H6BrNO
    MDL Number MFCD03095349
    CAS 13472-59-8
    Quantity 5g
    Flash Point 93°C (199°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 3-Bromo-2-methoxypyridine
    Assay 95%

    Description

    Applications
    It is employed in the facile synthesis of 2-unsubstituted benzofuran-3-carboxylates using diazo(trimethylsilyl)methyl magnesium bromide.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit