Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-2-methylpyridine, 97%

Manufacturer:  Alfa Aesar™ H2727103

Quantity 1g

 View more versions of this product

Catalog No. AAH2727103


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

38749-79-0
172.03
3-Bromo-2-picoline
C6H6BrN
MFCD00191224
3-Bromo-2-methylpyridine

Specifications

38749-79-0
172.03
3-Bromo-2-picoline
1g
3-Bromo-2-methylpyridine
C6H6BrN
MFCD00191224
76°C (17mmHg)
79°C (174°F)
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P301+P310-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit