Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-2-methylpyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
38749-79-0
C6H6BrN
172.02
AIPWPTPHMIYYOX-UHFFFAOYSA-N
3-bromo-2-picoline,2-methyl-3-bromopyridine,3-bromo-2-methyl-pyridine,pyridine, 3-bromo-2-methyl,2-methyl-3-bromo pyridine,pubchem1220,3-brom-2-methylpyridin,acmc-209j1t,3-bromo-2-methyl pyridine,ksc497m1l
1201003
3-bromo-2-methylpyridine
CC1=C(C=CC=N1)Br
38749-79-0
C6H6BrN
172.02
AIPWPTPHMIYYOX-UHFFFAOYSA-N
3-bromo-2-picoline,2-methyl-3-bromopyridine,3-bromo-2-methyl-pyridine,pyridine, 3-bromo-2-methyl,2-methyl-3-bromo pyridine,pubchem1220,3-brom-2-methylpyridin,acmc-209j1t,3-bromo-2-methyl pyridine,ksc497m1l
1201003
3-bromo-2-methylpyridine
CC1=C(C=CC=N1)Br
Flash Point 79°C
Boiling Point 65°C to 67°C (10.0 Torr)
Specific Gravity 1.495
Density 1.4950g/mL
Color Brown to Colorless
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 172.02
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-methylpyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger