Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-3-buten-1-ol, 97+%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
76334-36-6
C4H7BrO
151.003
MFCD00154041
RTKMFQOHBDVEBC-UHFFFAOYSA-N
3-bromo-3-buten-1-ol,3-buten-1-ol, 3-bromo,3bb,zlchem 79,acmc-20an90,3-bromanylbut-3-en-1-ol,3-bromo-3-buten-1-ol
533975
3-bromobut-3-en-1-ol
C=C(CCO)Br
76334-36-6
C4H7BrO
151.003
MFCD00154041
RTKMFQOHBDVEBC-UHFFFAOYSA-N
3-bromo-3-buten-1-ol,3-buten-1-ol, 3-bromo,3bb,zlchem 79,acmc-20an90,3-bromanylbut-3-en-1-ol,3-bromo-3-buten-1-ol
533975
3-bromobut-3-en-1-ol
C=C(CCO)Br
Density 1.522g/mL
Color Brown
Quantity 1g
Infrared Spectrum Authentic
Refractive Index 1.5000 to 1.5020 (20°C, 589nm)
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Linear Formula CH=C(Br)CHCHOH
Beilstein 01, IV, 2100
Formula Weight 151
Physical Form Liquid, Darkening On Exposure To Light
Percent Purity ≥97.0%
Chemical Name or Material 3-Bromo-3-buten-1-ol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.