Alfa Aesar™ 3-Bromo-3-buten-1-ol, 97%, stab. with copper

Manufacturer: Alfa Aesar™ H3411909

Quantity 10g

 View more versions of this product

Catalog No. AAH3411909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76334-36-6
    Molecular Formula C4H7BrO
    Formula Weight 151
    MDL Number MFCD00154041
    Chemical Name or Material 3-Bromo-3-buten-1-ol, stabilized with copper

    Specifications

    Formula Weight 151
    Boiling Point 64° to 65°C (9mmHg)
    Quantity 10g
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C4H7BrO
    MDL Number MFCD00154041
    CAS 76334-36-6
    Flash Point 110°C (230°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H318-H317
    Toxic if swallowed.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Recommended Storage 2° to 8°C
    Chemical Name or Material 3-Bromo-3-buten-1-ol, stabilized with copper
    Assay 97%

    Description

    Applications
    3-Bromo-3-buten-1-ol, 97%, stab. with copper is used as a building block for α-methylene lactones and tricyclic sesquiterpenes. It is used in the synthesis of 3-bromo-3-butenyl acrylate.

    Notes
    Light sensitive. Store in the dark. Store away from oxidizing agent, light.

    GHS H Statement
    H301-H318-H317
    Toxic if swallowed.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    2° to 8°C

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit