Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-4-chlorobenzotrifluoride, 97%

Click to view available options
Quantity:
5g
10g
50g
454-78-4
C7H3BrClF3
259.45
MFCD00018093
APSISOSWYXCEQX-UHFFFAOYSA-N
2-bromo-1-chloro-4-trifluoromethyl benzene,3-bromo-4-chlorobenzotrifluoride,benzene, 2-bromo-1-chloro-4-trifluoromethyl,3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene,4-chloro-3-bromobenzotrifluoride,3-bromo-4-chloro-a,a,a-trifluorotoluene,pubchem4837,ksc915a8j,apsisoswyxceqx-uhfffaoysa,3-bromo-4-chloro-benzotrifluoride
67988
2-bromo-1-chloro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)Cl
454-78-4
C7H3BrClF3
259.45
MFCD00018093
APSISOSWYXCEQX-UHFFFAOYSA-N
2-bromo-1-chloro-4-trifluoromethyl benzene,3-bromo-4-chlorobenzotrifluoride,benzene, 2-bromo-1-chloro-4-trifluoromethyl,3-bromo-4-chloro-alpha,alpha,alpha-trifluorotoluene,4-chloro-3-bromobenzotrifluoride,3-bromo-4-chloro-a,a,a-trifluorotoluene,pubchem4837,ksc915a8j,apsisoswyxceqx-uhfffaoysa,3-bromo-4-chloro-benzotrifluoride
67988
2-bromo-1-chloro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)Cl
Melting Point -23°C to -22°C
Boiling Point 191°C to 192°C
Flash Point 94°C (201°F)
Quantity 50g
Beilstein 1638470
Formula Weight 259.44
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-chlorobenzotrifluoride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning