Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-chloropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
36953-42-1
C5H3BrClN
192.44
MFCD00234008
QADXKWUCCGPQNR-UHFFFAOYSA-N
pyridine, 3-bromo-4-chloro,3-bromo-4-chloro-pyridine,4-chloro-3-bromopyridine,acmc-1ai1d,3-bromo-4-chloropyridine,,ksc222g5j,pyridine,3-bromo-4-chloro-,hydrochloride 1:1
817696
3-bromo-4-chloropyridine
C1=CN=CC(=C1Cl)Br
36953-42-1
C5H3BrClN
192.44
MFCD00234008
QADXKWUCCGPQNR-UHFFFAOYSA-N
pyridine, 3-bromo-4-chloro,3-bromo-4-chloro-pyridine,4-chloro-3-bromopyridine,acmc-1ai1d,3-bromo-4-chloropyridine,,ksc222g5j,pyridine,3-bromo-4-chloro-,hydrochloride 1:1
817696
3-bromo-4-chloropyridine
C1=CN=CC(=C1Cl)Br
Color Colorless to Yellow
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 192.44
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-chloropyridine
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning