Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-4-cyanopyridine, 95%

Manufacturer:  Alfa Aesar™ H27049MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27049MD


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

13958-98-0
183.01
3-Bromoisonicotinonitrile; 3-Bromopyridine-4-carbonitrile
C6H3BrN2
MFCD00234143
3-Bromo-4-cyanopyridine

Specifications

13958-98-0
183.01
92° to 94°C
250mg
3-Bromo-4-cyanopyridine
C6H3BrN2
MFCD00234143
3-Bromoisonicotinonitrile; 3-Bromopyridine-4-carbonitrile
Odorless
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit