Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-cyanopyridine, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
13958-98-0
C6H3BrN2
183.01
VPDIXJCBBUZNPO-UHFFFAOYSA-N
3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo
11074051
3-bromopyridine-4-carbonitrile
C1=CN=CC(=C1C#N)Br
13958-98-0
C6H3BrN2
183.01
VPDIXJCBBUZNPO-UHFFFAOYSA-N
3-bromo-4-cyanopyridine, 3-bromoisonicotinonitrile, 3-bromo-4-pyridinecarbonitrile, 4-pyridinecarbonitrile, 3-bromo, 3-bromo-isonicotinonitrile, pubchem17105, acmc-20a0fj, 5-bromoisonicotinonitrile, 4-pyridinecarbonitrile,3-bromo
11074051
3-bromopyridine-4-carbonitrile
C1=CN=CC(=C1C#N)Br
Quantity 5g
Melting Point 91°C to 94°C
Assay Percent Range 96% min. (GC)
Formula Weight 183.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-cyanopyridine

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS Signal Word: Danger

RUO – Research Use Only