Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3'-Bromo-4'-fluoroacetophenone, 98+%

Click to view available options
Quantity:
5g
25g
1007-15-4
C8H6BrFO
217.037
MFCD00042466
SZDWTGAORQQQGY-UHFFFAOYSA-N
3'-bromo-4'-fluoroacetophenone,3-bromo-4-fluoroacetophenone,1-3-bromo-4-fluorophenyl ethanone,1-3-bromo-4-fluorophenyl ethan-1-one,ethanone, 1-3-bromo-4-fluorophenyl,1-3-bromo-4-fluoro-phenyl-ethanone,1-acetyl-3-bromo-4-fluorobenzene,pubchem4100,acmc-1bol8,ksc499c8t
70508
1-(3-bromo-4-fluorophenyl)ethanone
CC(=O)C1=CC(=C(C=C1)F)Br
1007-15-4
C8H6BrFO
217.037
MFCD00042466
SZDWTGAORQQQGY-UHFFFAOYSA-N
3'-bromo-4'-fluoroacetophenone,3-bromo-4-fluoroacetophenone,1-3-bromo-4-fluorophenyl ethanone,1-3-bromo-4-fluorophenyl ethan-1-one,ethanone, 1-3-bromo-4-fluorophenyl,1-3-bromo-4-fluoro-phenyl-ethanone,1-acetyl-3-bromo-4-fluorobenzene,pubchem4100,acmc-1bol8,ksc499c8t
70508
1-(3-bromo-4-fluorophenyl)ethanone
CC(=O)C1=CC(=C(C=C1)F)Br
Flash Point >110°C (230°F)
Melting Point 55°C to 57°C
Boiling Point 150°C (12mmHg)
Quantity 25g
Beilstein 3088698
Formula Weight 217.04
Percent Purity ≥98%
Chemical Name or Material 3'-Bromo-4'-fluoroacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning