Alfa Aesar™ 3-Bromo-4-fluoroaniline, 98%

Manufacturer: Alfa Aesar™ B2561003

Quantity 1g

 View more versions of this product

Catalog No. AAB2561003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 656-64-4
    Molecular Formula C6H5BrFN
    Formula Weight 190.01
    MDL Number MFCD03788559
    UN Number UN2811
    Chemical Name or Material 3-Bromo-4-fluoroaniline

    Specifications

    Formula Weight 190.01
    Melting Point 34° to 36°C
    Boiling Point 235°C
    Quantity 1g
    Molecular Formula C6H5BrFN
    MDL Number MFCD03788559
    UN Number UN2811
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    CAS 656-64-4
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 3-Bromo-4-fluoroaniline
    Assay 98%

    Description

    Applications
    3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.

    Solubility
    Insoluble in water.

    Notes
    Incompatible with strong oxidizing agents, strong acids and bases.

    GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit