Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Bromo-4-fluoroaniline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
3-Bromo-4-fluoroaniline is used in organic synthesis as an intermediate of agrochemicals and active pharmaceutical ingredient.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
Melting Point | 34°C to 36°C |
Boiling Point | 235°C |
Quantity | 5 g |
UN Number | UN2811 |
Sensitivity | Air sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 190.01 |
Percent Purity | 98% |
Chemical Name or Material | 3-Bromo-4-fluoroaniline |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319 |
Hazard Statement | GHS H Statement H331-H302-H312-H315-H319 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction