Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-bromo-4-fluorobenzotrifluoride 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
68322-84-9
C7H3BrF4
242.999
MFCD00040938
RZJOIMPUMMQKFR-UHFFFAOYSA-N
3-bromo-4-fluorobenzotrifluoride,2-bromo-1-fluoro-4-trifluoromethyl benzene,2-fluoro-5-trifluoromethyl bromobenzene,3-bromo-alpha,alpha,alpha,4-tetrafluorotoluene,benzene, 2-bromo-1-fluoro-4-trifluoromethyl,1-bromo-2-fluoro-5-trifluoromethyl benzene,3-bromo-alpha,alpha,alpha-4-tetrafluorotoluene,pubchem1639,3-bromo-4-fluoro trifluoromethyl benzene
144295
2-bromo-1-fluoro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)F
68322-84-9
C7H3BrF4
242.999
MFCD00040938
RZJOIMPUMMQKFR-UHFFFAOYSA-N
3-bromo-4-fluorobenzotrifluoride,2-bromo-1-fluoro-4-trifluoromethyl benzene,2-fluoro-5-trifluoromethyl bromobenzene,3-bromo-alpha,alpha,alpha,4-tetrafluorotoluene,benzene, 2-bromo-1-fluoro-4-trifluoromethyl,1-bromo-2-fluoro-5-trifluoromethyl benzene,3-bromo-alpha,alpha,alpha-4-tetrafluorotoluene,pubchem1639,3-bromo-4-fluoro trifluoromethyl benzene
144295
2-bromo-1-fluoro-4-(trifluoromethyl)benzene
C1=CC(=C(C=C1C(F)(F)F)Br)F
Density 1.706g/mL
Packaging Glass bottle
Quantity 5g
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Infrared Spectrum Authentic
Color Colorless
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H3(F)CF3
Formula Weight 243
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 3-Bromo-4-fluorobenzotrifluoride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger