Alfa Aesar™ 3-Bromo-4-fluorobiphenyl, 98%

Manufacturer: Alfa Aesar™ B2531614

Quantity 25g

 View more versions of this product

Catalog No. AAB2531614

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 306935-88-6
    Molecular Formula C12H8BrF
    Formula Weight 251.1
    MDL Number MFCD01571091
    UN Number UN3151
    Chemical Name or Material 3-Bromo-4-fluorobiphenyl

    Specifications

    Formula Weight 251.1
    Quantity 25g
    Molecular Formula C12H8BrF
    MDL Number MFCD01571091
    CAS 306935-88-6
    UN Number UN3151
    DOT Information Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, LIQUID
    Chemical Name or Material 3-Bromo-4-fluorobiphenyl
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit