Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-4-fluorobiphenyl, 98%

Click to view available options
Quantity:
5g
25g
306935-88-6
C12H8BrF
251.098
MFCD01571091
COWXPZSVUXHAFS-UHFFFAOYSA-N
3-bromo-4-fluorobiphenyl,3-bromo-4-fluoro-1,1'-biphenyl,acmc-20an8u,1,1'-biphenyl,3-bromo-4-fluoro,2-bromanyl-1-fluoranyl-4-phenyl-benzene
2773367
2-bromo-1-fluoro-4-phenylbenzene
C1=CC=C(C=C1)C2=CC(=C(C=C2)F)Br
306935-88-6
C12H8BrF
251.098
MFCD01571091
COWXPZSVUXHAFS-UHFFFAOYSA-N
3-bromo-4-fluorobiphenyl,3-bromo-4-fluoro-1,1'-biphenyl,acmc-20an8u,1,1'-biphenyl,3-bromo-4-fluoro,2-bromanyl-1-fluoranyl-4-phenyl-benzene
2773367
2-bromo-1-fluoro-4-phenylbenzene
C1=CC=C(C=C1)C2=CC(=C(C=C2)F)Br
Quantity 5g
UN Number UN3151
Formula Weight 251.1
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-fluorobiphenyl

DOTInformation : Transport Hazard Class: 9; Packing Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, LIQUID