Alfa Aesar™ 3'-Bromo-4'-fluoropropiophenone, 98%

Manufacturer: Alfa Aesar™ B2542906

Quantity 5g

 View more versions of this product

Catalog No. AAB2542906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 202865-82-5
    Molecular Formula C9H8BrFO
    Formula Weight 231.06
    MDL Number MFCD00143012
    Chemical Name or Material 3'-Bromo-4'-fluoropropiophenone

    Specifications

    Formula Weight 231.06
    Molecular Formula C9H8BrFO
    MDL Number MFCD00143012
    Melting Point 60° to 64°C
    Quantity 5g
    CAS 202865-82-5
    Chemical Name or Material 3'-Bromo-4'-fluoropropiophenone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit