Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 3-Bromo-4-fluorotoluene, 99%
Click to view available options
Quantity:
5g
25g
Specifications
Density | 1.507 |
Boiling Point | 168°C to 170°C |
Refractive Index | 1.531 |
Quantity | 5g |
Flash Point | 73°C (163°F) |
Beilstein | 1680604 |
Formula Weight | 189.03 |
Percent Purity | 99% |
Chemical Name or Material | 3-Bromo-4-fluorotoluene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 207-201-4
TSCA : No
Recommended Storage : Ambient temperatures