Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-4-hydroxy-5-methoxycinnamic acid, 97%

Click to view available options
Quantity:
1g
5g
6948-33-0
C10H9BrO4
273.082
MFCD00017334
NGBVBJQUJJKRGF-NSCUHMNNSA-N
3-3-bromo-4-hydroxy-5-methoxyphenyl acrylic acid,5-bromoferulic acid,2e-3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid,3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid,3-bromo-4-hydroxy-5-methoxycinnamic acid,2e-3-3-bromo-4-hydroxy-5-methoxyphenyl acrylic acid,2-propenoic acid,3-3-bromo-4-hydroxy-5-methoxyphenyl,e-3-3-bromo-4-hydroxy-5-methoxy-phenyl prop-2-enoic acid,e-3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid
735789
(E)-3-(3-bromo-4-hydroxy-5-methoxyphenyl)prop-2-enoic acid
COC1=C(C(=CC(=C1)C=CC(=O)O)Br)O
6948-33-0
C10H9BrO4
273.082
MFCD00017334
NGBVBJQUJJKRGF-NSCUHMNNSA-N
3-3-bromo-4-hydroxy-5-methoxyphenyl acrylic acid,5-bromoferulic acid,2e-3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid,3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid,3-bromo-4-hydroxy-5-methoxycinnamic acid,2e-3-3-bromo-4-hydroxy-5-methoxyphenyl acrylic acid,2-propenoic acid,3-3-bromo-4-hydroxy-5-methoxyphenyl,e-3-3-bromo-4-hydroxy-5-methoxy-phenyl prop-2-enoic acid,e-3-3-bromo-4-hydroxy-5-methoxyphenyl prop-2-enoic acid
735789
(E)-3-(3-bromo-4-hydroxy-5-methoxyphenyl)prop-2-enoic acid
COC1=C(C(=CC(=C1)C=CC(=O)O)Br)O
Quantity 1g
Melting Point 255°C to 257°C
Odor Odorless
Formula Weight 273.09
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-hydroxy-5-methoxycinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures