Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methoxybenzonitrile, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
117572-79-9
C8H6BrNO
212.05
MFCD00143097
QHWZMDRKTYTPEE-UHFFFAOYSA-N
2-bromo-4-cyanoanisole,3-bromo-4-methoxylbenzonitrile,3-bromo-4-methoxybenzenecarbonitrile,3-bromo-4-methoxy-benzonitrile,benzonitrile, 3-bromo-4-methoxy,pubchem3795,acmc-1cexj,maybridge4_001971,3-bromo4-methoxybenzonitrile,ksc494m8l
736049
3-bromo-4-methoxybenzonitrile
COC1=C(C=C(C=C1)C#N)Br
117572-79-9
C8H6BrNO
212.05
MFCD00143097
QHWZMDRKTYTPEE-UHFFFAOYSA-N
2-bromo-4-cyanoanisole,3-bromo-4-methoxylbenzonitrile,3-bromo-4-methoxybenzenecarbonitrile,3-bromo-4-methoxy-benzonitrile,benzonitrile, 3-bromo-4-methoxy,pubchem3795,acmc-1cexj,maybridge4_001971,3-bromo4-methoxybenzonitrile,ksc494m8l
736049
3-bromo-4-methoxybenzonitrile
COC1=C(C=C(C=C1)C#N)Br
Quantity 10g
Melting Point 120.0°C to 121.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 212.05
Percent Purity 99%
Chemical Name or Material 3-Bromo-4-methoxybenzonitrile

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning