Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-4-methoxypyridine, 97%

Click to view available options
Quantity:
250mg
1g
5g
82257-09-8
C6H6BrNO
188.024
MFCD05664024
DSFJKFHIMBVWAV-UHFFFAOYSA-N
3-bromo-4-methoxy-pyridine,pyridine, 3-bromo-4-methoxy,pyridine,3-bromo-4-methoxy,pubchem6686,4-methoxy-3-bromopyridine,acmc-209po0,abbypharma ap-17-5958
642827
3-bromo-4-methoxypyridine
COC1=C(C=NC=C1)Br
82257-09-8
C6H6BrNO
188.024
MFCD05664024
DSFJKFHIMBVWAV-UHFFFAOYSA-N
3-bromo-4-methoxy-pyridine,pyridine, 3-bromo-4-methoxy,pyridine,3-bromo-4-methoxy,pubchem6686,4-methoxy-3-bromopyridine,acmc-209po0,abbypharma ap-17-5958
642827
3-bromo-4-methoxypyridine
COC1=C(C=NC=C1)Br
Applications
3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water (9.8 g/L).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Color Yellow
Quantity 5g
Beilstein 1423610
Formula Weight 188.02
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methoxypyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C