Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 3-Bromo-4-methylaniline, 97%

Click to view available options
Quantity:
5g
25g
7745-91-7
C7H8BrN
186.052
MFCD00134176
GRXMMIBZRMKADT-UHFFFAOYSA-N
3-bromo-p-toluidine,p-toluidine, 3-bromo,benzenamine, 3-bromo-4-methyl,3-bromo-4-methyl-phenylamine,3-bromo-1,4-toluidine,4-amino-2-bromotoluene,3-bromo-4-methylbenzenamine,3-bromo-4-methylphenylamine,3-bromo-para-toluidine,3-bromo-4-methyl-aniline
82187
3-bromo-4-methylaniline
CC1=C(C=C(C=C1)N)Br
7745-91-7
C7H8BrN
186.052
MFCD00134176
GRXMMIBZRMKADT-UHFFFAOYSA-N
3-bromo-p-toluidine,p-toluidine, 3-bromo,benzenamine, 3-bromo-4-methyl,3-bromo-4-methyl-phenylamine,3-bromo-1,4-toluidine,4-amino-2-bromotoluene,3-bromo-4-methylbenzenamine,3-bromo-4-methylphenylamine,3-bromo-para-toluidine,3-bromo-4-methyl-aniline
82187
3-bromo-4-methylaniline
CC1=C(C=C(C=C1)N)Br
Applications
3-Bromo-4-methylaniline is used in the synthesis of 1,7-dihalo Tröger′s base isomers. It is suitable for use to investigate the reactions of distonic 4-(N,N,N-trimethylammonium)-2-methylphenyl and 5-(N,N,N-trimethylammonium)-2-methylphenyl radical cations with O2 by ion-trap mass spectrometry.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Quantity 5g
Flash Point >110°C (230°F)
Melting Point 25°C to 28°C
Odor Characteristic
Boiling Point 254°C to 257°C
UN Number UN2811
Beilstein 1562057
Formula Weight 186.06
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methylaniline

GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : XU4375000