Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methylaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
7745-91-7
C7H8BrN
186.05
MFCD00134176
GRXMMIBZRMKADT-UHFFFAOYSA-N
3-bromo-p-toluidine,p-toluidine, 3-bromo,benzenamine, 3-bromo-4-methyl,3-bromo-4-methyl-phenylamine,3-bromo-1,4-toluidine,4-amino-2-bromotoluene,3-bromo-4-methylbenzenamine,3-bromo-4-methylphenylamine,3-bromo-para-toluidine,3-bromo-4-methyl-aniline
82187
3-bromo-4-methylaniline
CC1=C(C=C(C=C1)N)Br
7745-91-7
C7H8BrN
186.05
MFCD00134176
GRXMMIBZRMKADT-UHFFFAOYSA-N
3-bromo-p-toluidine,p-toluidine, 3-bromo,benzenamine, 3-bromo-4-methyl,3-bromo-4-methyl-phenylamine,3-bromo-1,4-toluidine,4-amino-2-bromotoluene,3-bromo-4-methylbenzenamine,3-bromo-4-methylphenylamine,3-bromo-para-toluidine,3-bromo-4-methyl-aniline
82187
3-bromo-4-methylaniline
CC1=C(C=C(C=C1)N)Br
Refractive Index 1.6110 to 1.613
Melting Point 27°C to 30°C
Boiling Point 254°C to 257°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Undesignated
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula BrC6H3(CH3)NH2
Formula Weight 186.05
Physical Form Liquid After Melting
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-methylaniline, 97%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-807-8