Alfa Aesar™ 3-Bromo-4-methylphenol, 98%

Manufacturer: Alfa Aesar™ H28689MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH28689MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 60710-39-6
    Molecular Formula C7H7BrO
    Formula Weight 187.04
    MDL Number MFCD08273793
    Synonym 3-Bromo-p-cresol
    Chemical Name or Material 3-Bromo-4-methylphenol

    Specifications

    Formula Weight 187.04
    Melting Point 54° to 57°C
    Quantity 250mg
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C7H7BrO
    MDL Number MFCD08273793
    Synonym 3-Bromo-p-cresol
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
    CAS 60710-39-6
    Chemical Name or Material 3-Bromo-4-methylphenol
    Assay 98%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

    Solubility
    Slightly soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit