Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-Bromo-4-methylthiophene, 95%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
Specifications
Flash Point | 78°C |
Boiling Point | 179°C to 183°C (770.0mmHg) |
Specific Gravity | 1.584 |
Refractive Index | 1.5770 to 1.579 |
Packaging | Glass bottle |
Density | 1.5840g/mL |
Infrared Spectrum | Authentic |
Color | Colorless to Yellow |
Quantity | 1g |
CAS Min % | 94.0 |
Show More |
Safety and Handling
GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning