Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Bromo-4-methylthiophene, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
30318-99-1
C5H5BrS
177.06
MFCD00130084
MBUSOPVRLCFJCS-UHFFFAOYSA-N
3-bromo-4-methyl-thiophene,thiophene, 3-bromo-4-methyl,pubchem21832,acmc-209her,3-bromo-4-methylthiophen,4-bromo-3-methylthiophene,bidd:gt0486,3-bromo-4-methylthiophene
2734935
3-bromo-4-methylthiophene
CC1=CSC=C1Br
30318-99-1
C5H5BrS
177.06
MFCD00130084
MBUSOPVRLCFJCS-UHFFFAOYSA-N
3-bromo-4-methyl-thiophene,thiophene, 3-bromo-4-methyl,pubchem21832,acmc-209her,3-bromo-4-methylthiophen,4-bromo-3-methylthiophene,bidd:gt0486,3-bromo-4-methylthiophene
2734935
3-bromo-4-methylthiophene
CC1=CSC=C1Br
Boiling Point 179°C to 183°C (770.0mmHg)
Flash Point 78°C
Specific Gravity 1.584
Refractive Index 1.5770 to 1.579
Density 1.5840g/mL
Color Colorless to Yellow
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 177.06
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 3-Bromo-4-methylthiophene
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning