Alfa Aesar™ 3-Bromo-5-chloro-2,6-dimethoxybenzamide, 97%

Manufacturer: Alfa Aesar™ B2473506

Quantity 5g

 View more versions of this product

Catalog No. AAB2473506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 175135-60-1
    Molecular Formula C9H9BrClNO3
    Formula Weight 294.53
    MDL Number MFCD00052965
    Chemical Name or Material 3-Bromo-5-chloro-2,6-dimethoxybenzamide

    Specifications

    Formula Weight 294.53
    Quantity 5g
    Molecular Formula C9H9BrClNO3
    MDL Number MFCD00052965
    CAS 175135-60-1
    Chemical Name or Material 3-Bromo-5-chloro-2,6-dimethoxybenzamide
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit